SY3H3D67E352 Synthonix Corporation

7-Fluoro-1,3-benzoxazole

  • Cas Number : 1267061-18-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/44543.html

Safety Information

MSDS https://synthonix.com/44543.html
Availability details for
Quantity
  • Availability
  • Add to Cart