SY3H3D67E359 Synthonix Corporation

4-Fluoro-3-hydroxybenzonitrile

  • Cas Number : 186590-04-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/44549.html

Safety Information

MSDS https://synthonix.com/44549.html
Availability details for
Quantity
  • Availability
  • Add to Cart