SY3H3D67E35B Synthonix Corporation

6-Fluoro-2(3H)-benzothiazolone

  • Cas Number : 63754-96-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/44553.html

Safety Information

MSDS https://synthonix.com/44553.html
Availability details for
Quantity
  • Availability
  • Add to Cart