SY3H3D67E35D Synthonix Corporation

4-Fluorobenzo[d]thiazol-2(3H)-one

  • Cas Number : 63754-97-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/44555.html

Safety Information

MSDS https://synthonix.com/44555.html
Availability details for
Quantity
  • Availability
  • Add to Cart