SY3H3D67E35E Synthonix Corporation

6-Fluorobenzo[d]isothiazol-3(2H)-one

  • Cas Number : 159803-11-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin USA
Product Link https://synthonix.com/44556.html

Safety Information

MSDS https://synthonix.com/44556.html
Availability details for
Quantity
  • Availability
  • Add to Cart