SY3H3D67E370 Synthonix Corporation

5-Fluoro-1H-pyrrolo[3,2-b]pyridine

  • MDL number MFCD08062552
  • Cas Number : 887570-96-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerator
Physical Form Liquid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/44799.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/44799.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart