SY3H3D67E38C Synthonix Corporation

3-Fluoro-4-methoxypicolinonitrile

  • Cas Number : 1427357-47-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/44843.html

Safety Information

MSDS https://synthonix.com/44843.html
Availability details for
Quantity
  • Availability
  • Add to Cart