SY3H3D67E394 Synthonix Corporation

Furo[3,4-b]pyridin-5(7H)-one

  • Cas Number : 5657-51-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/44926.html

Safety Information

MSDS https://synthonix.com/44926.html
Availability details for
Quantity
  • Availability
  • Add to Cart