SY3H3D67E3D0 Synthonix Corporation

3-(Furan-2-yl)acrylaldehyde

  • Cas Number : 623-30-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/45509.html

Safety Information

MSDS https://synthonix.com/45509.html
Availability details for
Quantity
  • Availability
  • Add to Cart