SY3H3D67E3DD Synthonix Corporation

2-Fluoro-N,4-dihydroxybenzimidamide

  • Cas Number : 880874-38-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/45669.html

Safety Information

MSDS https://synthonix.com/45669.html
Availability details for
Quantity
  • Availability
  • Add to Cart