SY3H3D67E3EA Synthonix Corporation

2-Fluoro-4-methylphenol

  • MDL number MFCD03094332
  • Cas Number : 452-81-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. refrigerator
Physical Form Light-yellow to yellow powder or crystal or liquid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/45958.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/45958.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart