SY3H3D67E416 Synthonix Corporation

(S)-1-(5-Fluoropyridin-3-yl)ethanamine hydrochloride

  • Cas Number : 1956436-47-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/46567.html

Safety Information

MSDS https://synthonix.com/46567.html
Availability details for
Quantity
  • Availability
  • Add to Cart