SY3H3D67E417 Synthonix Corporation

(S)-1-(6-Fluoropyridin-2-yl)ethanamine hydrochloride

  • MDL number MFCD28405348
  • Cas Number : 1956437-56-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Refrigerator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/46568.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/46568.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart