SY3H3D67E425 Synthonix Corporation

3-Fluoro-4-(methoxymethyl)aniline

  • Cas Number : 1341785-86-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/46814.html

Safety Information

MSDS https://synthonix.com/46814.html
Availability details for
Quantity
  • Availability
  • Add to Cart