SY3H3D67E426 Synthonix Corporation

(2-Fluoro-6-methoxyphenyl)methanamine

  • Cas Number : 150517-75-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/46815.html

Safety Information

MSDS https://synthonix.com/46815.html
Availability details for
Quantity
  • Availability
  • Add to Cart