SY3H3D67E42A Synthonix Corporation

4-Fluorophthalonitrile

  • Cas Number : 65610-14-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/47255.html

Safety Information

MSDS https://synthonix.com/47255.html
Availability details for
Quantity
  • Availability
  • Add to Cart