SY3H3D67E42D Synthonix Corporation

2-Fluoro-5-trifluoromethylbenzonitrile

  • MDL number MFCD00061280
  • Cas Number : 4088-84-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. refrigerator
Physical Form Clear liquid / Colour: Colorless - Very pale yellow
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/47279.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/47279.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart