SY3H3D67E44B Synthonix Corporation

2-Fluoro-5-(trifluoromethoxy)benzaldehyde

  • MDL number MFCD04115886
  • Cas Number : 886497-81-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Refrigerator
Physical Form Colorless to light-yellow liquid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/47488.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/47488.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart