SY3H3D67E455 Synthonix Corporation

7-Fluorobenzo[b]thiophene

  • Cas Number : 346592-74-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/47541.html

Safety Information

MSDS https://synthonix.com/47541.html
Availability details for
Quantity
  • Availability
  • Add to Cart