SY3H3D67E456 Synthonix Corporation

5-Fluoroquinazoline

  • MDL number MFCD18448849
  • Cas Number : 16499-43-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Refrigerator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/47554.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/47554.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart