SY3H3D67E45D Synthonix Corporation

2-(3-Fluoro-2-nitrophenyl)acetonitrile

  • Cas Number : 105003-88-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/47593.html

Safety Information

MSDS https://synthonix.com/47593.html
Availability details for
Quantity
  • Availability
  • Add to Cart