SY3H3D67E45E Synthonix Corporation

7-Fluoro-1H-indazole-3-carboxylic acid

  • MDL number MFCD07378920
  • Cas Number : 959236-59-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerator
Physical Form Pale-yellow to Yellow-brown Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/47594.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/47594.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart