SY3H3D67E468 Synthonix Corporation

3-Formyl-4-hydroxybenzonitrile

  • Cas Number : 74901-29-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/47714.html

Safety Information

MSDS https://synthonix.com/47714.html
Availability details for
Quantity
  • Availability
  • Add to Cart