SY3H3D67E483 Synthonix Corporation

6-Fluoro-2-methylbenzo[d]thiazole

  • MDL number MFCD00012319
  • Cas Number : 399-73-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. refrigerator
Physical Form Light-yellow to brown powder or crystals
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/47982.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/47982.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart