SY3H3D67E489 Synthonix Corporation

(5-Fluoro-2-nitrophenyl)acetic acid

  • MDL number MFCD07787601
  • Cas Number : 29640-98-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Refrigerator
Physical Form Crystal - Powder / Colour: White - Yellow
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/48051.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/48051.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart