SY3H3D67E48D Synthonix Corporation

4-Fluoro-2-methyl-3-nitrobenzoic acid

  • MDL number MFCD28797610
  • Cas Number : 1079991-68-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. refrigerator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/48055.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/48055.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart