SY3H3D67E491 Synthonix Corporation

4-Fluoroindolin-2-one

  • MDL number MFCD04112528
  • Cas Number : 138343-94-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Refrigerator
Physical Form Off white to Pale-yellow to Yellow-brown Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/48095.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/48095.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart