SY3H3D67E49C Synthonix Corporation

2-Fluoro-3-(hydroxymethyl)benzonitrile

  • MDL number MFCD18394036
  • Cas Number : 1261861-91-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Refrigerator
Physical Form White to off-white solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/48106.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/48106.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart