SY3H3D67E4A1 Synthonix Corporation

4-Fluoro-1H-indol-7-amine

  • Cas Number : 292636-13-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/48340.html

Safety Information

MSDS https://synthonix.com/48340.html
Availability details for
Quantity
  • Availability
  • Add to Cart