SY3H3D67E4C1 Synthonix Corporation

1-(2-Fluoro-6-hydroxyphenyl)ethanone

  • MDL number MFCD01090995
  • Cas Number : 93339-98-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerator
Physical Form Colorless to Pale-yellow to Yellow-brown Liquid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/48624.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/48624.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart