SY3H3D67E4CC Synthonix Corporation

2-(3-Fluorophenyl)-2-hydroxyacetic acid

  • MDL number MFCD03095383
  • Cas Number : 395-05-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/48687.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/48687.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart