SY3H3D67E508 Synthonix Corporation

(S)-1-(3-Fluorophenyl)ethylamine hydrochloride

  • Cas Number : 321429-48-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/49511.html

Safety Information

MSDS https://synthonix.com/49511.html
Availability details for
Quantity
  • Availability
  • Add to Cart