SY3H3D67E50A Synthonix Corporation

(2-Fluoro-6-methoxyphenyl)methanamine hydrochloride

  • Cas Number : 1214366-31-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/49513.html

Safety Information

MSDS https://synthonix.com/49513.html
Availability details for
Quantity
  • Availability
  • Add to Cart