SY3H3D67E513 Synthonix Corporation

1-Fluoro-4-(3-iodopropyl)benzene

  • Cas Number : 118156-84-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/49704.html

Safety Information

MSDS https://synthonix.com/49704.html
Availability details for
Quantity
  • Availability
  • Add to Cart