SY3H3D67E523 Synthonix Corporation

4-Fluoro-1H-indole-3-carbonitrile

  • Cas Number : 1260759-82-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/50672.html

Safety Information

MSDS https://synthonix.com/50672.html
Availability details for
Quantity
  • Availability
  • Add to Cart