SY3H3D67E52A Synthonix Corporation

3-Formyl-1H-indazole-5-carboxylic acid

  • MDL number MFCD06656900
  • Cas Number : 885519-98-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Refrigerator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/50827.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/50827.html
pictograms GHS07
hstatements H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart