SY3H3D67E530 Synthonix Corporation

3-Fluoroquinolin-5-ol

  • Cas Number : 1261729-67-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/50943.html

Safety Information

MSDS https://synthonix.com/50943.html
Availability details for
Quantity
  • Availability
  • Add to Cart