SY3H3D67E558 Synthonix Corporation

2-(3-Formylphenyl)acetic acid

  • Cas Number : 34956-29-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/51625.html

Safety Information

MSDS https://synthonix.com/51625.html
Availability details for
Quantity
  • Availability
  • Add to Cart