SY3H3D67E55D Synthonix Corporation

3-Fluoro-N-methoxy-N-methyl-4-nitrobenzamide

  • Cas Number : 863604-64-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/51692.html

Safety Information

MSDS https://synthonix.com/51692.html
Availability details for
Quantity
  • Availability
  • Add to Cart