SY3H3D67E55F Synthonix Corporation

2-(4-Fluorophenyl)propanoic acid

  • MDL number MFCD00054492
  • Cas Number : 75908-73-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/51933.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/51933.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart