SY3H3D67E566 Synthonix Corporation

1-(3-Fluoro-2-methoxyphenyl)ethanone

  • MDL number MFCD12545933
  • Cas Number : 295779-86-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerator
Physical Form Colorless to light-yellow liquid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/51947.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/51947.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart