SY3H3D67E576 Synthonix Corporation

5-Fluoro-2,3-dihydro-1H-inden-1-ol

  • Cas Number : 52085-92-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/52046.html

Safety Information

MSDS https://synthonix.com/52046.html
Availability details for
Quantity
  • Availability
  • Add to Cart