SY3H3D67E597 Synthonix Corporation

5-Fluoro-2,3-dimethyl-1H-indole

  • Cas Number : 526-47-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/52361.html

Safety Information

MSDS https://synthonix.com/52361.html
Availability details for
Quantity
  • Availability
  • Add to Cart