SY3H3D67E59B Synthonix Corporation

4-(4-Fluorophenyl)pyrrolidin-2-one

  • MDL number MFCD03447332
  • Cas Number : 264122-82-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. refrigerator
Physical Form White to Pale-yellow to Yellow-brown Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/52400.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/52400.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart