SY3H3D67E5A7 Synthonix Corporation

7-Fluoro-1,2,3,4-tetrahydronaphthalen-2-amine

  • MDL number MFCD06739166
  • Cas Number : 173998-63-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Refrigerator
Physical Form Liquid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/52752.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/52752.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart