SY3H3D67E5AB Synthonix Corporation

3-FLUORO-5-(ISOBUTOXY)BENZENEBORONICACID

  • MDL number MFCD07363783
  • Cas Number : 850589-57-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerator
Physical Form White to light-yellow powder or crystals
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/53102.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/53102.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart