SY3H3D67E5B3 Synthonix Corporation

3-(2-Fluoro-4-(trifluoromethyl)phenyl)acrylic acid

  • MDL number MFCD00236292
  • Cas Number : 262608-88-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Refrigerator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/53490.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/53490.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart