SY3H3D67E5C3 Synthonix Corporation

6-Fluorochroman-8-carbaldehyde

  • MDL number MFCD08236762
  • Cas Number : 82060-91-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. refrigerator
Physical Form Pale-yellow to Yellow-brown to Gray Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/54000.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/54000.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart