SY3H3D67E5C4 Synthonix Corporation

3-(3-Fluoro-4-methylphenyl)acrylic acid

  • MDL number MFCD01631413
  • Cas Number : 261951-72-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Refrigerator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/54001.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/54001.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart