SY3H3D67E5D6 Synthonix Corporation

(S)-3-((4-Fluorophenyl)thio)pyrrolidine hydrochloride

  • Cas Number : 1353995-35-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/54133.html

Safety Information

MSDS https://synthonix.com/54133.html
Availability details for
Quantity
  • Availability
  • Add to Cart